Learn More
Medchemexpress LLC Survodutide (TFA) | 2805997-46-8 | 99.9% | 4231.63 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYP41465MG
Encompass_Preferred
Survodutide is a potent, selective glucagon receptor/GLP-1 receptor (GCGR/GLP-1R) dual agonist. It is a 29-amino-acid peptide and a potent acylated peptide containing a C18 fatty acid, demonstrating robust anti-obesity efficacy by increasing energy expenditure and decreasing food intake.
  • Potent, selective glucagon receptor/GLP-1 receptor dual agonist
  • Demonstrates robust anti-obesity efficacy
  • Increases energy expenditure and decreases food intake
  • Enhanced proteolytic stability
  • Extended terminal half-life
Catalog No. 50-003-98534