Learn More
Med Vet International Oasis Chromic Gut Cassette, Size 1, Length of 50M, Each
Supplier:  Med Vet International CG01
Encompass_Preferred
The veterinary use Oasis Chromic Gut suture cassette is a natural absorbable suture that has a brown thread color and is extremely strong tensile strength, designed to provide secure knotting, and be more pliable and flexible. Size: 1. Length: 50M.
Catalog No. 50-264-3579