Learn More
Med Vet International Ethicon PDS II Suture, Size 2-0, FS-1, 27in, 12/Box
Supplier:  Med Vet International Z451G
Encompass_Preferred
The Ethicon Z451G suture is a high-quality absorbable Polydioxanone surgical suture. It measures 27in. in length, has a size of 2-0, they are dyed to provide greater visibility while suturing, and typically have an absorption rate of 182-238 days. This suture comes with a FS-1 reverse cutting needle and has 12 sutures in a box.
Catalog No. 50-252-2109