Learn More
Med Vet International Ethicon Ethilon Nylon Suture, Size 1, CTX, 30in, Black, 36/Box
Supplier:  Med Vet International 830H
Encompass_Preferred
The Ethicon 830H suture is a high-quality non-absorbable Ethilon Nylon suture. It measures 30in in length, has a size of 1, and are dyed black to provide greater visibility while suturing. This suture comes with a CTX 48mm 1/2c Taper Needle and has 36 sutures in a box.
Catalog No. 50-252-2509