Learn More
Med Vet International Oasis Monofyl Suture, Size 2-0, with NFS-1 Needle, 12/Box, Veterinary Use Only
Supplier:  Med Vet International MVY943V
Encompass_Preferred
The Oasis veterinary use Monofyl suture with needle is a monofilament synthetic absorbable suture that is comparable to Ethicon™ Monocryl®. The Monofyl suture has an absorption rate between 91 and 119 days. Size: 2-0. Needle Type: NFS-1, 24mm 3/8 Circle Reverse Cutting. Length: 30in. Quantity: 12/box
Catalog No. NC1905232