Learn More
Med Vet International Oasis Nylon Suture Cassette, Size 2-0, Length of 100M, Each
Supplier:  Med Vet International MF20
Encompass_Preferred
The veterinary use Oasis Nylon suturecassette is a monofilament non-absorbable suture. This Nylon suture has extremely high tensile strength that exceeds USP requirements. It provides excellent knot security, while the smooth surface allows for easy passage through the tissue. The thread color of this suture is blue and has a composition of polyamide. Size: 2-0. Length: 100M.
Catalog No. 50-264-3562