Learn More
Med Vet International Ethicon Suture PDS II 4-0 FS-2 36/Box Z422H
Supplier:  Med Vet International Z422H
Encompass_Preferred
The Ethicon Z422H suture is a high-quality absorbable Polydioxanone surgical suture. It measures 27in. in length, has a size of 4-0, they are dyed undyed to provide greater visibility while suturing, and typically have an absorption rate of 182-238 days. This suture comes with a FS-2 reverse cutting needle and has 36 sutures in a box.
Catalog No. NC1985436