Learn More
Med Vet International Ethicon Vicryl Suture, Size 2-0, CT-1, 36in, 36/Box
Supplier:  Med Vet International J345H
Encompass_Preferred
The Ethicon J345H suture is a high-quality absorbable Polyglactin 910 surgical suture. It measures 36in. in length, has a size of 2-0, they are dyed violet to provide greater visibility while suturing, and typically have an absorption rate of 56-70 days. This suture comes with a CT-1 taper point needle and has 36 sutures in a box.
Catalog No. 50-118-0819