Learn More
Med Vet International Ethicon Vicryl Polyglactin 910 Plus Antibacterial Suture, Size 5-0, Ps-5, 18in, Undyed, 12/Box
Supplier:  Med Vet International VCP593G
Encompass_Preferred
The Ethicon VCP593G suture is a high-quality absorbable Vicryl Polyglactin 910 Plus Antibacterial suture. It measures 18in in length, has a size of 5-0, and are undyed. This suture comes with a PS-5 13mm 1/2c Cutting Edge Prime Reverse Needle and has 12 sutures in a box. Primary code substitute for discontinued code J593G
Catalog No. 50-283-0027