Learn More
Med Vet International Ethicon Vicryl Polyglactin 910 Suture, Size 5-0, PC-3, 18in, Undyed, 12/Box
Supplier:  Med Vet International J844G
Encompass_Preferred
The Ethicon J844G suture is a high-quality absorbable Vicryl Polyglactin 910 suture. It measures 18in in length, has a size of 5-0, and are undyed. This suture comes with a PC-3 16mm 3/8c Conventional Cutting Needle and has 12 sutures in a box.
Catalog No. 50-252-2347