Learn More
Med Vet International Ethicon Chromic Gut Suture, Size 0, CT, 27in, 36/Box
Supplier:  Med Vet International 802H
Encompass_Preferred
The Ethicon 802H suture is a high-quality absorbable Chromic Gut surgical suture. It measures 27in. in length, has a size of 0, they are dyed brown to provide greater visibility while suturing, and typically have an absorption rate of 90 days. This suture comes with a CT taper point needle and has 36 sutures in a box.
Catalog No. 50-209-2831