Learn More
Med Vet International Dynarex Braided Black Silk Sutures, Non Absorbable, Black, 3-0, C6 Needle, 18", 12/Box
Supplier:  Med Vet International 9143
Encompass_Preferred
Used to hold body tissues together after an injury or surgery. Application generally involves using a needle with an attached length of thread. A non-absorbable suture made from natural silk that is degummed to remove extraneous materials Braided for strength and easy handling Available in various thread thickness and needle dimensions to accommodate a variety of procedures Sterile
Catalog No. 50-283-0279