Learn More
Med Vet International Ethicon Vicryl Suture, Size 4-0, FS-1, 27in, 36/Box
Supplier:  Med Vet International J451H
Encompass_Preferred
The Ethicon J451H suture is a high-quality absorbable Polyglactin 910 surgical suture. It measures 27in. in length, has a size of 4-0, they are dyed violet to provide greater visibility while suturing, and typically have an absorption rate of 56-70 days. This suture comes with a FS-1 reverse cutting needle and has 36 sutures in a box.
Catalog No. 50-209-2882