Learn More
Medchemexpress LLC SUVN-911 | 2414674-71-6 | 99.13% | 261.15 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY136146100MG
Encompass_Preferred
SUVN-911 is a potent, selective, brain penetrated and orally bioavailable neuronal nicotinic acetylcholine α4β2 receptor antagonist, with a Ki of 1.5 nM. SUVN-911 has antidepressant activity.
  • Potent, selective, brain-penetrant, orally bioavailable α4β2 receptor antagonist (Ki: 1.5 nM).
  • Demonstrates antidepressant activity.
  • High selectivity for α4β2 over α3β4 nAChR.
  • Good selectivity against 70+ receptors (GPCRs, ion channels, enzymes, etc.).
  • Devoid of cardiovascular and gastrointestinal side effects.
  • Significant antidepressant effects in male Wistar rats (1-10 mg/kg, p.o., daily, 3 days).
  • Shows metabolic stability in rats.
  • High oral exposure, longer half-life, and adequate brain penetration in Wistar rats (3 mg/kg, p.o.).
Catalog No. 50-003-49772