Learn More
Medchemexpress LLC Suvorexant impurity 1 | 2941-79-9 | ≥98.0% | C16H14O4 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY78572100MG
Encompass_Preferred
Suvorexant impurity 1 is an impurity of Suvorexant, presented as an off-white to brown solid.
  • Research reagent
  • Not for medical, clinical diagnostic, or food use in humans or animals
  • Powder storage: -20°C for 3 years or 4°C for 2 years
  • In-solvent storage: -80°C for 6 months or -20°C for 1 month
Catalog No. 50-004-34441