Learn More
Medchemexpress LLC Suzetrigine phenol | 2649467-91-2 | 99.4% | 459.37 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY16090310MM/1ML
Encompass_Preferred
Suzetrigine phenol is the phenolate form of the sodium channel modulator Suzetrigine (HY-148800). Suzetrigine (VX-548) is an orally active and highly selective inhibitor of NaV1.8. It is a highly selective, orally active sodium channel modulator for research purposes.
  • Sodium channel modulator
  • Orally active
  • Highly selective inhibitor of NaV1.8
  • Appearance: solid, white to off-white
  • For research use only
Catalog No. 50-003-92628