Learn More
Medchemexpress LLC SVMP Crotalus adama 10ug

Supplier: Medchemexpress LLC HYP7145510UG
Recombinant snake venom metalloproteinase (SVMP) derived from Crotalus adamanteus, supplied lyophilized with an N-terminal 6xHis tag. Expressed in Escherichia coli and provided in small research quantities, the protein is suitable for biochemical assays investigating metalloproteinase activity and serpin interactions; molecular sequence and storage recommendations are supplied by the manufacturer.
- Recombinant protein expressed in E. coli with an N-terminal 6xHis tag.
- Purity greater than 90% by reducing SDS-PAGE.
- Lyophilized formulation from 10 mM Tris-HCl, 1 mM EDTA, 6% trehalose, pH 8.0.
- Molecular weight approximately 27.1 kDa.
- Sequence provided for verification and experimental design.
- Store at -20°C; after reconstitution, stable at 4°C for one week or frozen for long-term storage.
- Supplied in small research sizes suitable for biochemical assays and interaction studies.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.