Learn More
Medchemexpress LLC SW-100 100mg | 2126744-35-0 | 316.78 g·mol⁻¹ | C17H17ClN2O2 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY115475100MG
Encompass_Preferred
SW-100 is a selective histone deacetylase 6 (HDAC6) inhibitor for research use, reported with an IC50 of 2.3 nM. It displays at least 1,000-fold selectivity versus other HDAC isozymes and has improved blood-brain barrier penetration, supporting its use in neurological disease studies.
  • Potent HDAC6 inhibition with IC50 of 2.3 nM.
  • At least 1,000-fold selectivity versus other HDAC isozymes.
  • High purity suitable for research applications.
  • Available in common laboratory pack sizes, including 100 mg.
  • Soluble in DMSO for solution preparation and storage.
Catalog No. 50-002-28161