Learn More
Medchemexpress LLC SW-100 50mg | 2126744-35-0 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY11547550MG
Encompass_Preferred
SW-100 is a selective histone deacetylase 6 (HDAC6) inhibitor used as a research reagent in biochemical and cellular studies. It is highly potent and provided in solid and solution formats for in vitro and in vivo applications.
  • Selective HDAC6 inhibition with reported IC50 = 2.3 nM.
  • High purity appropriate for research use.
  • Soluble in DMSO for in vitro assays.
  • Formulations available for in vivo dosing studies.
  • Stable under recommended storage conditions.
Catalog No. 50-002-28163