Learn More
Beckman Coulter Bucket set for SW 32.1 rotor - 17 mL tubes, set of 6, includes caps and O-rings
Supplier:  Beckman Coulter 369693
Encompass
Replacement bucket assembly designed for swing-bucket ultracentrifuge rotors. Each set includes six buckets with caps and O-rings and is sized to hold 17 mL tubes, providing secure tube support during high-speed runs.
  • Set of six replacement buckets
  • Includes caps and O-rings
  • Holds 17 mL tubes
  • Compatible with SW 32 series swing-bucket rotors
  • Intended for replacing worn or damaged buckets in swing-bucket rotors
Catalog No. NC2878876
Request a Quote
To request a quote for this item, please select Request a Quote to reach our dedicated support team.