Learn More
Apexbio Technology LLC SW033291, 25mg. Cas: 459147-39-8 MFCD: MFCD02973087. 15-PGDH enzyme inhibitor
SDP
Supplier:  Apexbio Technology LLC A870925
Encompass_Preferred
SW033291 is a potent inhibitor of 15-PGDH enzyme with IC50 value of 1.5 nM and dissociation constant Ki app value of 0.1 nM. SW033291 inhibits 15-PGDH with noncompetitive pattern. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5180