Learn More
Medchemexpress LLC HY-124778 5mg Medchemexpress, SW106065 CAS:62289-81-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1247785MG
Encompass_Preferred
Medchemexpress, HY-124778 5mg SW106065 CAS:62289-81-0 SW106065 is an apoptosis inducer in malignant peripheral nerve sheath tumors (MPNST). SW106065 inhibits ATP consumption of sMPNST and other models of MPNST with an EC50 of 1 µM. SW106065 can be used for MPNST research. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1050