Learn More
IDEX Health & Science Upchurch Swaging Tool used to work with TinyTight Fittings
Supplier:  IDEX Health & Science Upchurch M150
Encompass_Preferred
This Swaging tool is used to work with TinyTight Fittings. Use this tool by placing in a vice, then tighten tubing, fitting, and ferrule into the tool as you would into any port. Once removed, the swaged ferrule will be held in place on the tubing.
Catalog No. NC3236682