Learn More
ABclonal Technology SWAP70 Rabbit pAb
SDP
Supplier:  ABclonal Technology A148571000UL
Encompass_Preferred
Enables cadherin binding activity. Predicted to be involved in regulation of actin polymerization or depolymerization. Predicted to act upstream of or within isotype switching. Located in actin cytoskeleton, cytoplasm, and plasma membrane.
Catalog No. 50-001-08380