Learn More
Cayman Chemical SWAY100135hydrochLRIde 10mg
SDP
Supplier:  Cayman Chemical 3285110 MG
Encompass_Preferred
A 5-HT1A antagonist (IC50 =15.5 nM in a radioligand binding assay); selective for 5-HT1A over 5-HT1B, 5-HT1C, and 5-HT2 receptors, as well as dopamine D1 and D2, and α1-, α2-, and β-adrenergic receptors (IC50s = >1 µM for all); increases the number of entries into and percentage of time spent in the open arms of the elevated plus maze in mice at 10 mg/kg.
Catalog No. 50-295-0796