Learn More
Cayman Chemical Swerta bmaculta Swertsn 5mg
SDP
Supplier:  Cayman Chemical 347445 MG
Encompass_Preferred
A flavonoid C-glycoside with diverse biological activities; inhibits SGLT2 in HEK293 at 7.5 UG/ml; is an adenosine A1 receptor antagonist (IC50 = 137 µM); inhibits HBV replication in HepG2 2.2.15 cells at 0.2-5 µM; decreases blood glucose levels in a mouse model of streptozotocin-induced diabetes; prevents scopolamine-induced increases in escape latency in the Morris water maze in mice
Catalog No. 50-291-0263