Learn More
Medchemexpress LLC Swertisin | 6991-10-2 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYN218910MM/1ML
Encompass_Preferred
Swertisin is an oral adenosine A1 receptor antagonist and an SGLT2 inhibitor. It exhibits anti-diabetic and antioxidant properties, inhibits HBV, and has been shown to improve cognitive and memory impairments in mice.
  • Oral adenosine A1 receptor antagonist
  • SGLT2 inhibitor
  • Exhibits anti-diabetic properties
  • Exhibits antioxidant properties
  • Inhibits HBV
  • Improves cognitive and memory impairments in mice
Catalog No. 50-004-20623