Learn More
Kingfisher Biotech Inc Swine IFN gamma Recombinant Protein (Yeast, Endotoxin - Free) - 25 ug
SDP
Supplier:  Kingfisher Biotech Inc RP0126S025
Encompass
The Swine IFN gamma yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Swine IFN gamma applications are for cell culture, ELISA standard, and Western Blot Control. The Swine IFN gamma yeast-derived recombinant protein can be purchased in multiple sizes. Swine IFN gamma Specifications: (Molecular Weight: 16.8 kDa) (Amino Acid Sequence: QAPFFKEITI LKDYFNASTS DVPNGGPLFL EILKNWKEES DKKIIQSQIV SFYFKFFEIF KDNQAIQRSM DVIKQDMFQR FLNGSSGKLN DFEKLIKIPV DNLQIQRKAI SELIKVMNDL SPRSNLRKRK RSQTMFQGQR ASK) (Gene ID: 396991). For research use only. Made in the USA
Catalog No. 50-241-3688