Learn More
World Precision Instrument Swiss High Precision Rounded Tip Cutting Tweezers, 11.5 cm
SDP
Supplier:  World Precision Instrument 504745
Encompass
WPI's Swiss Cutting Tweezers, also known as cutting forceps or scissors forceps, are designed for cutting tissues or other materials during surgical procedures. Cutting forceps are ideal for use in general surgery, orthopedic surgery, ophthalmology, and more. The primary purpose of cutting forceps is to provide surgeons with a precise and controlled tool for cutting tissues, sutures, or other materials in the operating room. These are hand forceps with a spring action letting you pinch the blades together to control the amount of pressure used. These forceps have sharp, cutting edges that allow for clean and accurate incisions. These cutting forceps have angled blades predominantly for surgical applications. They are also handy in working with electronics, because you can quickly snip soft copper, gold, silver, and stainless steel wire or braids. They are 11.5 cm (4.5") long. Choose either surgical grade stainless steel or ones with tungsten carbide inserts.
Catalog No. 50-253-9298