Learn More
MSC Swiss Precision Instrument Inc Plug & Pin Gage, Class ZZ Minus, 0.286 in Diameter, Steel
Supplier:  MSC 78374824
Encompass
This plug and pin gage is a precision instrument designed for accurate measurement applications. It features a Class ZZ Minus tolerance, making it ideal for verifying bore and hole sizes with high accuracy.
  • Diameter of 0.2860 in
  • Constructed from steel
  • Tolerance of 0.000200 in
Catalog No. 50-005-27251