Learn More
MSC Swiss Precision Instruments Plug Gage, Class ZZ, Minus, Steel, 0.225 inch
Supplier: MSC 78374212
This plug gage is a precision measuring tool designed for accurately gaging hole sizes and slot widths. It features a Class ZZ minus tolerance of 0.0002 inch, ensuring precise measurements for quality control and Go/No-Go applications. Constructed from durable steel, this gage is suitable for various measurement tasks in manufacturing and quality assurance.
- Precision tool for measuring hole sizes and slot widths.
- Ideal for quick and accurate measurements.
- Suitable for NO-GO gaging inspection.
- Constructed from durable steel.
- Features a Class ZZ minus tolerance of 0.0002 "
- Includes a certificate of accuracy traceable to NIST.
- Equipped with a pin gage holder.