Learn More
Perkin Elmer US LLC Swivel Tip for High Pressure Clamp
Supplier:  Perkin Elmer US LLC L1272324
Encompass_Preferred
Successful ATR analysis depends on how well the sample being analyzed comes into contact with the crystal. The High Pressure Clamp ensures the perfect amount of pressure. The clamp is shipped with three different tips: flat, concave, and swivel. Each tip is designed for specific types of samples.
Catalog No. 50-211-3526