Learn More
Medchemexpress LLC SYBR green I (chloride) | 2225748-05-8 | 98.1% | 545.18 | C32H37ClN4S | 10 MG

Supplier: Medchemexpress LLC HYD119110MG
SYBR green I chloride is a DNA-binding fluorescent dye supplied as a solid powder for nucleic acid staining and detection in molecular biology assays. It offers high purity and convenient solubility in DMSO for preparation of concentrated stock solutions.
- Nucleic acid fluorescent dye that binds preferentially to double-stranded DNA.
- High purity: 98.1%.
- Molecular formula C32H37ClN4S; molecular weight 545.18.
- Soluble in DMSO at 50 mg/mL; may require ultrasonic agitation.
- Store product at 4°C, sealed and protected from moisture and light.
- Recommended stock solution storage: -80°C for up to 6 months, -20°C for up to 1 month.
- Supplied in small research quantities suitable for laboratory use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.