Learn More
Cayman Chemical Syk InhIbItr II 1mg
SDP
Supplier:  Cayman Chemical 182021 MG
Encompass_Preferred
A cell-permeable, selective, and reversible inhibitor of Syk (IC50 = 41 nM); prevents FcεRI-mediated 5-HT release in RBL-2H3 cells in vitro (IC50 = 460 nM) and inhibits passive cutaneous anaphylaxis reactions in mice (ID50 = 13.2 mg/kg, s.c.)
Catalog No. 50-296-5729