Learn More
Ellsworth Adhesive Co SYLGARD 184 SILICONE 3.9KG
Supplier: Ellsworth Adhesive Co DC2065622
2-part, 10:1 mix, transparent encapsulant with good flame resistance. 10:1 mix ratio; flowable; room temperature or heat cure; high tensile strength; same as Sylgard 182 Elastomer but with RT cure capability; UL recognized; Mil Spec approved.