Learn More
TARGETMOL CHEMICALS INC SYM 2081 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T169635MG
Encompass_Preferred
Also available in 1 mg, 10 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes.SYM 2081 is a specific kainate receptor agonist with an IC50 value of 35 nM for [3H]-kainate binding. SYM 2081 depolarizes the muscle and reduces the EPSP amplitude. Purity 99.59%
Catalog No. 50-287-8828