Learn More
Waters Corp Waters Symmetry C18 Column, 5 μm Particle, 100 Å Pore, 4.6 mm ID, 50 mm Length
Supplier:  Waters Corp 186000207
Encompass_Preferred
Symmetry C18 columns from Waters are engineered to deliver highly reproducible chromatographic results. These columns utilize monofunctionally bonded C18 ligands on high-purity, base-deactivated silica, adhering to the industry's tightest specifications. Manufactured in certified facilities, each column ensures meticulous quality control and consistent performance for HPLC systems.
  • Achieves highly reproducible chromatographic results with tight specifications and high-purity, base-deactivated silica C18 ligands.
  • Minimizes resolution variations and ensures repeatable batch-to-batch results through narrow ligand surface coverage.
  • Avoids labor and lab costs by eliminating revalidation or new procedure building per batch.
  • Manufactured in cGMP, ISO9001, ISO13485 certified facilities for reliable results.
Catalog No. 50-819-043