Learn More
Waters Corp Symmetry C18 Column, 100Å, 3.5 μm, 1 mm x 50 mm
Supplier:  Waters Corp WAT106056
Encompass_Preferred
Symmetry C18 columns are silica-based HPLC columns designed for reproducible and rugged performance. They are manufactured using high-purity silica and tightly controlled processes, ensuring consistent column-to-column and batch-to-batch reproducibility.
  • General-purpose, high-efficiency reversed-phase column.
  • Delivers maximum reproducibility for analytical methods.
  • Utilizes C18 chemistry.
  • Suitable for a wide range of applications, including pharmaceuticals, environmental analysis, food and beverage, and clinical research.
Catalog No. 50-786-025