Learn More
Waters Corp Waters Column, C18, 3.5 μm, 2.1 mm ID, 100 mm Length
Supplier:  Waters Corp WAT058965
Encompass_Preferred
Symmetry C18 columns utilize monofunctionally bonded C18 ligands on a high purity, base-deactivated silica. They offer highly reproducible chromatographic outcomes by meeting tight industry specifications, reducing the need for method revalidation. Manufacturing is cGMP, ISO9001, and ISO13485 certified, ensuring consistent HPLC column performance.
  • Engineered with high purity raw materials and precise column packing procedures for reproducibility.
  • Narrowest ligand surface coverage specifications in the industry, leading to minimal shifts in resolution and more reproducible batch-to-batch results.
  • Facilitates easier method transfer between laboratories globally.
  • Delivered with a three-page Certificate of Analysis.
  • Operating temperature range of 20°C to 45°C provides enhanced selectivity, lower solvent viscosity, and increased mass transfer rates.
Catalog No. 50-785-857