Learn More
Waters Corp WATERS Symmetry C18 Column, 100Å, 3.5 μm, 3 mm x 100 mm
Supplier:  Waters Corp 186000696
Encompass_Preferred
Symmetry C18 columns feature monofunctionally bonded C18 ligands on a high purity base-deactivated silica. These columns are engineered for highly reproducible chromatographic results due to their tight specifications and consistent ligand surface coverage. Each column includes a three-page Certificate of Analysis.
  • Features monofunctionally bonded C18 ligands on a high purity base-deactivated silica.
  • Offers unparalleled reproducibility.
  • Features tightest ligand surface coverage standards.
  • Leads to fewer variations in resolution and consistent batch-to-batch results.
  • Engineered with high purity raw materials and column packing techniques.
  • Accompanied by a three-page Certificate of Analysis detailing specifications and outcomes of 22 crucial tests.
  • Allows for labor cost savings by reducing the need for re-validation and new method creation.
Catalog No. 50-780-462