Learn More
Waters Corp Waters C18 Column, 100Å, 3.5 μm, 3 mm × 150 mm
Supplier:  Waters Corp 186000695
Encompass_Preferred
These C18 columns feature monofunctionally bonded C18 ligands on a high purity base-deactivated silica, offering the tightest specifications in the industry for highly reproducible chromatographic results.
  • Monofunctionally bonded C18 ligands on high purity base-deactivated silica.
  • Industry’s tightest column specifications for highly reproducible results.
  • Delivers consistent reproducibility, reducing method revalidation.
  • Manufactured in cGMP, ISO9001, and ISO13485 certified facilities.
  • Produced with high purity raw materials and precise packaging for reproducible HPLC.
  • Narrow ligand density specifications for minimal resolution shifts and reproducible batch-to-batch results.
  • Shipped with a three-page Certificate of Analysis detailing specifications and 22 critical tests.
Catalog No. 50-780-461