Learn More
Waters Corp Waters Corporation Column, C18, 100Å, 3.5 μm, 3 mm x 50 mm
Supplier:  Waters Corp 186002612
Encompass_Preferred
Symmetry C18 columns are engineered for high efficiency and accuracy in HPLC. Utilizing monofunctionally bonded C18 ligands on high purity base-deactivated silica, these columns deliver unparalleled reproducibility and consistent results, adhering to stringent industry standards for ligand surface coverage.
  • Monofunctionally bonded C18 ligands on high purity base-deactivated silica.
  • C18 chemistry, reversed phase separation.
  • Particle size of 3.5 μm; pore size of 100 Å.
  • 3 mm inner diameter, 50 mm length.
  • pH range of 2 to 8; max pressure 6000 psi (415 Bar).
  • Exceptional reproducibility and consistent batch results.
  • Includes a three-page certificate of analysis.
  • Manufactured in cGMP, ISO9001, and ISO13485 certified facilities.
Catalog No. 50-781-241