Learn More
Waters Corp Symmetry C18 Column, 100Å, 3.5 µm, 4.6 mm X 30 mm, 1/pk
Supplier:  Waters Corp 186000271
Encompass_Preferred
Symmetry C18 columns feature monofunctionally bonded C18 ligands on a high purity base-deactived silica. Symmetry columns have the tightest column specifications in the industry giving resulting highly reproducible chromatographic results.
Catalog No. 50-780-159