Learn More
Waters Corp Symmetry C18 Column, 300 Å, 3.5 μm, 1 mm x 150 mm
Supplier: Waters Corp 186000185
Symmetry columns are designed for long-term compliance in HPLC methods, ensuring reliable peak shape, selectivity, and reproducibility. This particular C18 column supports legacy methods and offers precise separation capabilities.
- C18 chemistry for reversed phase separation
- pH range of 2 to 8
- Maximum pressure of 6000 psi (400 bar)
- Particle size of 3.5 μm
- Pore size of 300 Å
- Optimized for HPLC systems
- Inner diameter of 1 mm
- Length of 150 mm
- USP Classification L1
- Suitable for peptide applications
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.