Learn More
Waters Corp Waters Columns, C18, 5 μm, 4.6 mm, 150 mm
Supplier: Waters Corp WAT045905
Symmetry C18 columns feature monofunctionally bonded C18 ligands on a high-purity base-deactivated silica. Designed with the tightest column specifications, they ensure highly reproducible chromatographic results, exceeding standards for HPLC column performance.
- Unmatched column-to-column and batch-to-batch reproducibility.
- Offers reliable, rugged, and robust methods, increasing productivity and easing method transfer.
- Minimizes resolution shifts for reproducible results with narrow ligand surface coverage.
- Enables rapid analysis and purification, reducing run time by 50%.
- Guarantees consistent performance for up to 10,000 injections due to optimized stability.
- Versatile for separating non-polar to moderately polar compounds in various analysis fields.