Learn More
Waters Corp Symmetry Intelligent Speed Column, C18, 5 μm, 100 Å, 2.1 mm x 20 mm
Supplier:  Waters Corp 186002070
Encompass_Preferred
Symmetry C18 columns are designed with monofunctionally bonded C18 ligands on a high purity base-deactivated silica substrate. These columns are characterized by having the most stringent column specifications within the industry, which contributes to achieving highly reproducible chromatographic results. The column has a particle size of 5 μm, a pore size of 100 Å, an inner diameter of 2.1 mm, and a length of 20 mm. It operates within a pH range of 2 to 8 and can withstand a maximum pressure of 6000 psi (415 Bar). It is endcapped, exhibits low silanol activity, and features a spherical particle shape. It is formatted as an Intelligent Speed (IS) Column for HPLC systems.
  • Features monofunctionally bonded C18 ligands on a high purity base-deactivated silica.
  • Possesses the tightest column specifications in the industry.
  • Results in highly reproducible chromatographic results.
Catalog No. 50-780-926