Learn More
Waters Corp Waters Symmetry C8 Column, 100Å, 5 μm, 4.6 mm X 250 mm
Supplier:  Waters Corp WAT054270
Encompass_Preferred
Symmetry C8 columns are designed for High-Performance Liquid Chromatography (HPLC) applications, offering exceptional reproducibility. Featuring monofunctionally bonded C8 ligands on high purity base-deactivated silica, they ensure consistent results and easier method transfer. These columns allow method optimization at neutral or low pH without compromising peak shape.
  • Monofunctionally bonded C8 ligands on high purity base-deactivated silica.
  • Designed for rugged and reproducible performance across a pH range of 2–8.
  • Facilitates easier method transfer between labs globally.
  • Includes a three-page certificate of analysis with 22 critical tests.
  • Enables rapid analysis and purification for high-throughput lead generation.
  • Decreases run times by 50% without sacrificing resolution.
Catalog No. 50-818-758