Learn More
Waters Corp Waters C8 Column, 100Å, 3.5 μm, 2.1 mm X 100 mm
Supplier:  Waters Corp WAT058961
Encompass_Preferred
These columns feature monofunctionally bonded C8 ligands on a high purity base-deactivated silica, offering tight industry specifications for highly reproducible chromatographic results.
  • Tightest column specifications in the industry, resulting in highly reproducible chromatographic results.
  • Unsurpassed column-to-column and batch-to-batch reproducibility.
  • Achieves optimal repeatability and superior peak form.
  • Manufactured in cGMP, ISO9001, and ISO13485 certified facilities, ensuring strict control and adherence to industry requirements.
  • Delivers consistent batch-to-batch results with minimal resolution variance and exceptional repeatability year over year.
  • Compatible with guards, vials, and elution plates.
Catalog No. 50-785-856