Learn More
Waters Corp VanGuard Cartridge, 100Å, 3.5 μm, 2.1 x 5 mm, C8
Supplier:  Waters Corp 186007733
Encompass_Preferred
Symmetry C8 VanGuard Cartridges extend analytical column lifetime and performance by removing particulate contamination from the mobile phase stream. This cartridge is optimized to protect all 2.1 mm and 3.0 mm I.D. analytical columns containing 3.5 μm sorbent particles. It acts as a barrier between the injector and the analytical column, improving efficiency by obstructing the entry of chemical impurities and suspended particles.
  • Made from the same packing material as C8 columns.
  • Manufactured in a cGMP, ISO 9001-certified facility for quality and durability.
  • Each packing material batch is chromatographically tested to high standards.
  • Equipment tested for peak performance, enhancing selectivity, sensitivity, and speed.
  • High-performance packing materials ensure long-term preservation.
  • Designed for biological fluid cleanup and temperature control.
Catalog No. 50-239-1337