Learn More
Waters Corp Symmetry Shield RP8 Column, 100 à , 3.5 µm, 4.6 mm X 75 mm, 1/pk, with eConnect Column Tag
Supplier:  Waters Corp WAT094263RF
Encompass_Preferred
Symmetry Shield RP8 Column, 100 à , 3.5 µm, 4.6 mm X 75 mm, 1/pk, with eConnect Column Tag
Catalog No. 50-239-1815